Learn More
Thermo Scientific™ BetaSil™ C18 Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
€ 507.42
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | |||||
11548720
|
Thermo Scientific™
70105-012106 |
2.1 mm | 10 mm |
€ 507.42
Pack of 4 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
11553231
|
Thermo Scientific™
70105-022106 |
2.1 mm | 20 mm |
€ 507.42
Pack of 4 |
In Stock
Log in to see stock. |
|||||
Description
Offer convenient, easy to use protection for analytical HPLC columns
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Ligand on 100Å silica
- High surface area with high bonded phase coverage
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Specifications
C18 |
Product Title
Thermo Scientific™ BetaSil™ C18 Javelin™ Guard Columns