Learn More
JULABO™ Thermal H300 Bath Fluid
Thermal H300 is a transparent, high-performance, hydrocarbon-based heat transfer fluid that has very low toxicity.
Brand: JULABO™ 8940113
This item is not returnable.
View return policy
Description
- Broad temperature range
- High stability
- Heat conductivity
- Odorless
- Long fluid life
Specifications
5 L | |
40°C to 300°C | |
None |
Forte HAT | |
Bath Fluid |