Learn More
Julabo™ Thermal Bath Fluids
Click to view available options
Color:
Clear
Light Brown
Light Yellow
Transparent
Quantity:
5 L
10 L
Model:
H10
H20S
H250S
H350
H5
HS
HY
Thermal G
Thermal M
Temperature Range:
-20°C to +180°C
-30°C to +80°C
-40°C to +180°C
-50°C to +105°C
-80°C to +55°C
0°C to 220°C
20°C to 250°C
40°C to 170°C
50°C to 350°C
Unit Size:
10 L
5 L
Pack of 5
Description
- For closed systems like the Presto or Forte
- Has wide temperature ranges
- Low viscosity
- High stability
- Good heat conductivity
- Minimum odor
- Low corrosion tendency
- Low toxicity
- Long shelf life
Specifications
Specifications
| Color | Transparent |
| Temperature Range | 40°C to 170°C |
| Type | Thermal Bath Fluid |
| Fluid Delivery Type | Thermal Bath |
| For Use With (Application) | Ideal for temperature control applications and guarantee safe and reliable operation, viscosity, oxi |
| Includes | Drain tap |
| Model | Thermal M |
| Quantity | 10 L |
| Temperature (Metric) Maximum | >255°C |
| Viscosity | 293mm2/s at 20°C |