missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium for AAS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 25.95 - € 56.20
Chemical Identifiers
CAS | 7440-28-0 |
---|---|
Molecular Formula | Tl |
Molecular Weight (g/mol) | 204.38 |
MDL Number | MFCD00134063 |
InChI Key | BKVIYDNLLOSFOA-UHFFFAOYSA-N |
PubChem CID | 5359464 |
ChEBI | CHEBI:30440 |
IUPAC Name | thallium |
SMILES | [Tl] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12963295
|
Fisher Chemical
B 161 100 |
100 mL | Bottle |
€ 25.95
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12973295
|
Fisher Chemical
B 161 500 |
500 mL | Bottle |
€ 56.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7440-28-0 | |
204.38 | |
BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
CHEBI:30440 | |
[Tl] |
Tl | |
MFCD00134063 | |
5359464 | |
thallium |
Specifications
1g/L ±0.002g/L | |
Flame Single-element Standard Solution | |
MFCD00134063 | |
[Tl] | |
204.38 | |
CHEBI:30440 | |
Liquid |
7440-28-0 | |
Tl | |
BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
thallium | |
5359464 | |
2% HNO3 | |
Thallium |
Safety and Handling
H302+H332
P261;P264;P270;P271;P301+P312;P304+P340;P330;P501
Warning
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Thallium (Tl) 1000mg/l in HNO3 2%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .