Learn More
Texwipe™ TOC Cleaning Validation Kits
Shop All Texwipe Products
Click to view available options
Product Type:
Validation Kit
Validation Kit, Long Handle Swab
For Use With (Application):
Sampling 12 Production Areas
Sampling 72 Production Areas
Unit Size:
18 sets
Each
Pack of 18
Beschreibung
Beinhaltet
Vials, swabs, and labels for sampling 12 or 72 production areas, and container for sample transport.
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
- Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
- Prewashed vials have one-piece cap and septa design for easy sampling and analysis
- Polyethylene overcap protects septum surface from contamination until time of analysis
- Sloped shoulder design minimizes headspace for reduced background TOC level
- Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
- Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
- Double layered, double knit polyester head entraps contaminants in knit structure during sampling
- Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
- Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
- 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Zertifizierungen
Certificate of Analysis included on each box.
Spezifikation
Spezifikation
| Product Type | Validation Kit |
| For Use With (Application) | Sampling 72 Production Areas |
| Includes | 72 40 mL clear vials bonded septa cap, 144 Low TOC Alpha™ Sampling Swab, 72 blank vial labels |
| Quantity | 1/Cs. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur