Learn More
WTW™ TetraCon™ Conductivity Electrode 325/C
WTW carries two special measuring cells, which are specifically geared for these applications: The TetraCon™ S consists of an electrode body, which is perfectly fitted to measure pasteous samples thanks to its shovel shape and is easy to clean. The TetraCon™ 325/C is fitted with a chemical-resistant head made of PEEK to measure especially acidic samples.
Brand:  WTW™ 301900
798.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 10786253
  • € 913.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Specifications
0.475 cm-1
TetraCon™ Conductivity Electrode 325/C
Temperature
Acidic environments
1.5 m
Integrated temperature probe
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ TetraCon™ Conductivity Electrode 325/C > Cell Constant: 0.475cm��