missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenazine-d6, MedChemExpress

Product Code. 16482239
Encompass
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16482239

Brand: Medchem Express HYB0590S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Tetrabenazine D6 is the deuterium labeled Tetrabenazine, which is a VMAT-inhibitor used for treatment of hyperkinetic movement disorder.

Tetrabenazine D6 is the deuterium labeled Tetrabenazine, which is a VMAT-inhibitor used for treatment of hyperkinetic movement disorder.

Specifications

Chemical Name or Material Tetrabenazine-d6
CAS 1392826-25-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C19H21D6NO3
Color Off-White
Synonym Ro 1-9569-d6
SMILES O=C1[C@H](CC(C)C)CN2CCC3=CC(OC([2H])([2H])[2H])=C(OC([2H])([2H])[2H])C=C3[C@@]2([H])C1
Industry Type Neuroscience-Neuromodulation
Molecular Weight (g/mol) 323.46
Quantity 1 mg
Formula Weight 323.46
Percent Purity 98.3%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.