Learn More
Testosterone-16,16,17-d3 3-phenylpropionate, Honeywell Fluka™
Product Code. 15940262 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
CAS 876054-55-6
Packaging Glass Bottle
Quantity 10 mg
Grade Analytical Standard
For Use With (Application) Steroids and Hormones
Physical Form Solid