Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, Without Rim, Medium Wall
Product Code. p-200009477 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
5 mL
12 mL
13 mL
17 mL
21 mL
27 mL
32 mL
34 mL
53 mL
57 mL
112 mL
Length (Metric):
75 mm
100 mm
125 mm
130 mm
150 mm
160 mm
180 mm
200 mm
Diameter (Metric):
12 mm
14 mm
16 mm
18 mm
20 mm
24 mm
25 mm
30 mm
Förpackningsstorlek
Pack of 100
Pack of 50
Produktkode 17981142
Kun null tilbage
Tilføj til indkøbskurv
Edge
Tilføj til indkøbskurv
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from borosilicate glass 3.3
  • Thick-walled and therefore mechanically very resistant
  • With round bottom
TRUSTED_SUSTAINABILITY
Tekniske data
Capacity (Metric) 5 mL
Length (Metric) 75 mm
Material Borosilicate Glass (3.3)
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Diameter (Metric) 12 mm
Product Type Test Tube
Wall Thickness 1.0 mm
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.