Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Shop All Thermo Scientific Products
Click to view available options
Color:
Blue
Green
Orange
Red
White
Holds:
Tubes (1.5 mL)
Tubes (10 to 13 mm)
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Tubes (17 to 20 mm)
Tubes (21 to 25 mm)
Tubes (26 to 30 mm)
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Array:
3 x 3
3 x 8 array
4 x 10
4 x 4
4 x 5
4 x 6
6 x 12
6 x 6
8 x 12
Unit Size:
Each
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Specifications
| Product Type | Test Tube Rack |
| Description | Green; 26 to 30 mm; 3 x 8 array |
| Array | 3 x 8 array |
| Color | Green |
| Holds | Tubes (26 to 30 mm) |
| Positions | 24 |
| For Use With (Equipment) | MaxQ™ Shaker Platform |