Learn More
Julabo™ Test Tube Rack
Test tube rack made of high grade plastic or stainless steel
Brand:  Julabo™ 9970303
 View more versions of this product
Product Code. 15470632
  • € 68.35 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Test tube racks for water baths
High grade plastic
30 mm dia.
Up to +100°C
21
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Julabo™ Test Tube Rack > No. of Positions: 21