Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1kg
5kg
Description
- Proprietary formulation
- Use 50 capsules per liter
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ FDA |
| Content And Storage | 15°C to 30°C |
| Format | Capsule |
| Grade | Molecular biology |
| Product Type | Microbiology Media |
| Formulation | Yeast extract, Tryptone broth, Sodium Chloride and Vitamins |
| For Use With (Application) | For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |
| Quantity | 1000 g |