Learn More
Liebisch™ Evaporator with Fixed Heating Block
Metal block thermostat combined with a patented multiplex valve depot for superior evaporation operations. Liebisch™ Evaporator with Fixed Heating Block can evaporate various samples under targeted aeration.
Brand: Liebisch™ 56006101
Description
- Metal block thermostat / height-adjustable multiplex valve cepot
- Interchangeable stainless steel luer needles with plug-in connections.
- valve depot can be swiveled sideways for convenient sample loading
Specifications
230 V | |
Ambient to 200°C | |
260 x 180 x 135 mm |