Learn More
Abnova™ TERF2 293T Cell Transient Overexpression Lysate (Denatured)
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | TERF2 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC024890 |
| Molecular Weight (g/mol) | Theoretical M.W. 28.3kDa |
| Plasmid | pCMV-TERF2 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-TERF2 antibody (Mfr. No. H00007014-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |