Learn More
Oakton™ Temperature Controllers
Provide excellent control, accuracy, and power capabilities.
€ 1500.00
Specifications
For Use With (Application) Pilot process plants, R and D labs and OEM requirements
Product Type Temperature Controller
For Use With (Equipment) J, K, T, E, N, B, R and S probes
View More Specs
Products 1
Product Code Brand Includes Model Price Quantity & Availability  
Product Code Brand Includes Model Price Quantity & Availability  
11766508
View Documents
Oakton™
68900-13
Meter Only Advanced, 230V
€ 1500.00
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
Description
Description
Ideal for pilot process plants, R & D labs, and OEM requirements.
Probes
  • Controllers accept J, K, T, E, N, B, R, and S thermocouple-type probes with miniconnectors Advanced controller also accepts 400 Series thermistor probes with 1/4 in. (6.3mm) phono plug and RTD probes with 3-pin connectors
Field calibration offset
    Improves accuracy by compensating for individual probe error Resolution is 0.1°C below 999.9°C and 1°C above 999.9°C Accuracy: for Type J, K, T, E, and N probes, ±0.1% of reading, ±0.4°C (±0.7°F) above -100°C (-248°F) or ±0.1% of reading, ±1°C (±1.8°F) below -100°C (-248°F); for Type R, S and B probes, ±0.1% of reading, ±1°C (±1.8°F); for thermistor and RTD probes, ±0.1% of reading, ±0.4°C (±0.7°F)
Programmable
    Two-line alphanumeric display of set-up and operational parameters Set temperatures using Celsius, Fahrenheit, Kelvin, or Rankine scales Alarms are fully programmable with adjustable hysteresis and audible alarm option Choose from alarms off; manual reset; or automatic reset Process alarms trigger if temperature goes outside a set range and can be set for HIGH, LOW, or HIGH/LOW modes Deviation alarms trigger if temperature deviates from a single setpoint and can be set for HIGH, LOW, HIGH/LOW, or BAND modes Advanced controller includes SPDT alarm relay to set a circuit to either open or close when triggered
Safety Features
  • Fail-safe operation with adjustable overtemperature shutdown and loop break cutoff Output power to load device automatically shuts down if sensor or control loop breaks If overtemperature setpoint is exceeded, system shuts down automatically Output power receptacle fused separately for added safety Nonvolatile memory protects set-up parameters and saves parameters if power is lost
Standard Controller
  • Maximum heater output is 115/230V, 10A, 1150/2300w Select from ON/OFF, AUTOTUNE for standard proportional integral derivative (PID) applications
  • Programmable PID for nonstandard PID applications
Advanced Controller
  • All the standard controller's features plus higher heater output capacity of 115/230V, 15A, 1725/3450w, up to nine programmable ramp and soak profiles, bidirectional RS-232-C computer interface, and a Windows™ software program† Features an isolated 4 to 20mA output, scalable for positive or negative slope, 250ohm resistor for conversion to a 1 to 5V output signal, and an output connector
Order Info
Controllers are available in 115V (with 6 ft. long power cord) or 230V (with a European-style power cord set and power output connector). Dimensions: 10L x 7 3/8W x 3 3/4 in.H (25 x 18.5 x 9.4cm).
Specifications
Specifications
Pilot process plants, R and D labs and OEM requirements
Temperature Controller
J, K, T, E, N, B, R and S probes