Learn More
BINDER™ Temperature Calibration Certificate
Product Code. 30238267 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO Series
KB Series
KBF PRO Series
KBF Series
Calibration Points:
15 to 18 Measuring Points
27 Measuring Points
28 Measuring Points
29 Measuring Points
30 Measuring Points
9 Measuring Points
Unit Size:
Each
Product Code. 30238267
Brand: BINDER™ 80122426
Temperature measurement and certificate, measured at specified temperature.
  • Calibration measurement taken at specified temperature.
  • Choose between 9, 15 to 18, 27, 28, 29, or 30 measurement points
Specifications
For Use With (Equipment) KBF Series
Calibration Points 29 Measuring Points
Duration 1 Day
Product Type Calibration Certificate