Learn More
Kinesis™ TELOS Filtration Column
Brand:  Kinesis™ 910-0000-009T
 View more versions of this product
Product Code. 15156793
  • € 280.00 / Pack of 100
Estimated Shipment: 29-07-2025
to see stock.
Explore more special offers
Description
  • 5μm
  • PTFE
  • Specifications
    Specifications
    6 mL
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Kinesis™ TELOS Filtration Column > 6mL