Learn More
RETSCH Teflon Grinding Jars
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For MM 200
For MM 301 Mixer mill
Capacity (Metric):
10 mL
25 mL
Unit Size:
Each
Specifications
Specifications
| Product Type | Grinding Jar |
| Capacity (Metric) | 25 mL |
| For Use With (Equipment) | For MM 301 Mixer mill |
| Material | Teflon |