Learn More
Juchheim Laborgeraete™ Teclu Safety Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
Brand: Juchheim Laborgeraete™ 01262 DE
This item is not returnable.
View return policy
Description
- For operation with option of natural gas or propane gas
- With needle valve
- With DVGW certification
- With air control screen
- Particularly good stand due to heavy, high-quality material
Specifications
Bunsen Burner | |
Brass | |
Needle Valve |
Natural Gas | |
Gas consumption approx. 116 L/h (0.0813 kg/h), Heating capacity approx. 3398 kJ/h |