Learn More
Technico™ Pocket Refractometer
Click to view available options
Scale Range:
0 to 10 Brix
0 to 100 Salinity
0 to 15 Brix
28 to 62 Brix
Unit Size:
Each
Specifications
Specifications
| Type | Pocket Refractometer |
| Scale Range | 0 to 15 Brix |
| Refractive Index | 1.333 to 1.356 |
| Model | DIGIT-015 |
| Calibration | Zero calibration with distilled water at 20°C |
| For Use With (Application) | Measuring low sugar concentrations and low refractive values |
| Weight (Metric) | 160 g |
| Dimensions (L x W x H) | 55 x 80 x 205 mm |