Learn More
Medline™ Technico™ DIGIT-025 Sugar Pocket refractometer
Brand:  Medline™ 8100.0250M
Product Code. 10004592
  • € 186.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Specifications
Wine Pocket Refractometer
DIGIT-025
1 Each
0 to 25% Brix
Zero calibration with distilled water at 20°