missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCS PIM-1 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
ATP-competitive Pim-1 kinase inhibitor that displays selectivity over Pim-2 and MEK1/2 (IC50 values are 50, > 20000 and > 20000 nM for Pim-1, Pim-2 and MEK1/2 respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 367.2 |
Percent Purity | >98% |
Chemical Name or Material | TCS PIM-1 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'