Learn More
MP Biomedicals™ Taq Pol Bulk
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 250U
1000U
250U
3 x 250U
5 x 1000U
Unit Size:
1000 units
250 units
750 units
Description
- Ready for CE IVD labeling of clinical applications
Specifications
Specifications
| Concentration | 5 U/μL |
| Description | Taq Pol Bulk |
| Quantity | 10 x 250U |
| Includes | Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
For Resaerch Use Only