Learn More
Kartell™ Tapered Hose Coupling
Polyethylene disconnectors provide a quick, flexible, and safe alternative to expensive adapters. Kartell™ Tapered Hose Couplings tightly join glass, rubber, or plastic hoses together.
Brand: Kartell™ 7043600
8.58 EUR valid until 2025-01-17
Use promo code "21645" to get your promotional price.
Description
- Useful for joining hoses of glass, rubber or plastic
- Connector fits tightly together
- Replaces expensive adapters
Specifications
CE/IVD | |
Bore Size 3.6 mm | |
8/9/10 mm | |
Glass, rubber, or plastic hoses | |
3.6 mm |
8 to 10 mm | |
Polyethylene | |
Hose Coupling | |
100 | |
Natural |