missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ AADvanced™ Dead Cell Stain Kit

SYTOX™ AADvanced™ Dead Cell Stain is a new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry.

Brand:  Molecular Probes™ S10349

122.61 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10287064

  • € 183.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry
  • The dye is spectrally similar to 7-AAD but with rapid kinetics of uptake and relatively low CVs
  • SYTOX™ AADvanced™ Dead Cell Stain penetrates cells more efficiently than 7-AAD providing better separation of live and dead cells
  • It can also be used with fixed cells for DNA content analysis when paired with RNAse treatment
  • Kits contain a vial(s) of dried dye and anhydrous DMSO providing stable shelf life
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Impermeant
Contains 1 vial of SYTOX™ AADvanced™ dead cell stain and 200 μL dimethylsulfoxide (DMSO).

Store at ≤-20°C and protect from light.
Fluorescence
Flow Cytometer
SYTOX™ AADvanced™
546 nm
Tube(s)
100 Reactions
DMSO (Dimethylsulfoxide)
Red
SYTOX™ AADvanced™ Dead Cell Stain Kit
Viability Assay
Dead Cell Stain Kit
647 nm
Solid
Molecular Probes™, SYTOX™
Room Temperature
Nucleus, Nucleic Acids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2866034 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog NumberS10349
Lot Number2626248 Certificate TypeCertificate of Analysis Date09/08/2023 Catalog NumberS10349
Lot Number2480140 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog NumberS10349
Lot Number2356813 Certificate TypeCertificate of Analysis Date23/07/2021 Catalog NumberS10349
Lot Number2291522 Certificate TypeCertificate of Analysis Date09/03/2021 Catalog NumberS10349
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ SYTOX™ AADvanced™ Dead Cell Stain Kit > 100 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.