missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lotz Lagertechnik™ Swivel Stool with Leather Seat
€ 155.00
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12613255
|
Lotz Lagertechnik™
3861.1-04 |
Red |
€ 155.00
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
12653255
|
Lotz Lagertechnik™
3861.1-10 |
Black |
€ 155.00
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Specifications
540 to 720 mm | |
360 mm |