Learn More
SVL™ Cone/Screw Thread Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
Brand:  SVL 670-83
 View more versions of this product
Product Code. 15597740
Description
Description
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVL™ Screw Thread and Quickfit™ Ground Glass Cone
85 mm
29/32
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SVL™ Cone/Screw Thread Adapter > Cone size: 29/32; Joint size: 30