Learn More
SVL™ Cone/Screw Thread Adapter
Click to view available options
Cone Size:
14/23
24/29
Unit Size:
Pack of 10
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
Specifications
| Cone Size | 14/23 |
| For Use With (Equipment) | SVL™ and Quickfit™ Glassware |
| Material | Borosilicate Glass |
| Product Type | Cone/Screw thread adapter |
| Standard Taper Joint No. | 15 |