Learn More
Quickfit™ Claisen Still Head Adapter
Click to view available options
Unit Size:
Each
Pack of 5
Joint Type:
Condenser: 19/26, Flask: 24/29
Condenser: 24/29, Flask: 24/29
Size:
Socket: 14/23
Description
Similar to the SH4 still heads but with extra socket to accept dropping funnel or stirrer etc.
Specifications
Specifications
| Cone Size | 14/23 |
| For Use With (Equipment) | Used for thermometer, dropping funnel, stirrer |
| Material | Borosilicate Glass |
| Product Type | Claisen still head Adapter |