Learn More
Invitrogen™ SureCast™ Gel Handcast Station
Leak-free, confident handcasting of your polyacrylamide gels with the SureCast Handcast System.
Brand:  Invitrogen™ HC1000
80.62 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Product Code. 15519404
  • € 96.75 / Each
Explore more special offers
Description
Description
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
Pour your own polyacrylamide gels easily and confidently with the SureCast Handcast System and SureCast Handcast Reagents. The system features durable glass plates that are securely sealed using a single motion load-and-lock mechanism. The unique tilt feature of the SureCast Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.
Recommended Storage
Room Temperature
Services
Up to 12 months
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SureCast Handcast System
SureCast
1 station
Mini
SureCast Gel Handcast Station
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ SureCast™ Gel Handcast Station > 1 Casting Station
For Research Use Only. Not for use in diagnostic procedures.