Learn More
Cytiva Superdex™ Peptide 3.2/300 Prepacked Precision Column
For high-resolution gel filtration of peptides and small biomolecules with molecular weights between 100 and 7000 in micropreparative scale
Brand:  Cytiva 29036231
Product Code. 15107966
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Prepacked with Superdex Peptide media
  • Spherical composite of cross-linked agarose and dextran
  • Supplied with two fingertight 1/16 in. male connectors for connection
  • For sensitive and high resolution gel filtration of proteins, peptides, polynucleotides and other biomolecules
  • Used for polishing step in purification process
  • With new hardware design for direct connection to ÄKTATM systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 to 0.15 mL/min.
13 to 15 μm
Superdex Pre-packed Column
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Superdex Peptide, 3.2/303
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
SDS
Documents
Documents
Certificates
Special Offers
Special Offers