Learn More
Cytiva Superdex™ 75 3.2/300 Prepacked Precision Column
For sensitive and high-resolution gel filtration separations of biomolecules in micropreparative scale
Brand:  Cytiva 29036230
Product Code. 15117966
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Prepacked with Superdex 75 media
  • For molecules with molecular weight between 3,000 and 70,000
  • Spherical composite of cross-linked agarose and dextran
  • Supplied with two fingertight 1/16 in. male connectors for connection
  • For polishing step in purification process
  • With new hardware design for direct connection to ÄATKAM systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 to 0.10 mL/min.
Superdex Pre-packed Column
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers