missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Sulphate Cuvette Tests
€ 154.00
Specifications
No. of Tests | 25 |
---|---|
Storage Requirements | 15°C to 25°C |
Test Method | Barium Sulphate |
Type | Sulphate Cuvette Test |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10686213
|
Hach Lange™
LCK353 |
150 to 900mg/L SO4 |
€ 154.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
10192014
|
Hach Lange™
LCK153 |
40 to 150mg/L SO4 |
€ 154.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Specifications
25 | |
15°C to 25°C | |
Barium Sulphate | |
Sulphate Cuvette Test |
Safety and Handling
missing translation for 'shelfLife' : 36 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfat/Sulphate/Sulfate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : Glycerol, n-propanol, Hydrochloric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title