missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, Thermo Scientific Chemicals Product Code.: 10484755

Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, Thermo Scientific Chemicals

Product Code. 10484755
5 mL, Glass Bottle
Click to view available options
Quantity:
5 mL
50 mL
100 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10484755

Brand: Thermo Scientific Acros 175480050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-19-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 100.08
MDL Number MFCD00003542
InChI Key QAOWNCQODCNURD-ZSJDYOACSA-N
Synonym sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic
PubChem CID 160992
IUPAC Name dideuterio sulfate
SMILES [2H]OS(=O)(=O)O[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.8700g/mL
Deuterium (D) 99.50% min.
Packaging Glass Bottle
Quantity 5 mL
Linear Formula D2SO4
Formula Weight 100.09
Percent Purity 97.5 to 102.5%
Grade NMR
Chemical Name or Material Sulfuric acid-d2, 99.5+ atom % D
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464514 Certificate TypeCertificate of Analysis Date24/07/2024 Catalog Number175480500,175480050,175481000
Lot NumberA0433660 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number175480500,175480050,175481000
Lot NumberA0449304 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number175480500,175480050,175481000
Lot NumberA0450302 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number175480500,175480050,175481000
Lot NumberA0395225 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number175480500,175480050,175481000
compliance-icons
Product Identifier
  • Sulfuric acid-d2, 98 wt.% in D2O, 99.5+ atom % D
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.