missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+ atom % D
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
50 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
50mL
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Density | 1.8700g/mL |
| Deuterium (D) | 99.50% min. |
| Packaging | Glass Bottle |
| Quantity | 50 mL |
| Linear Formula | D2SO4 |
| Formula Weight | 100.09 |
| Percent Purity | 97.5 to 102.5% |
| Grade | NMR |
| Show More |
Safety and Handling
Product Identifier
- Sulfuric acid-d2, 98 wt.% in D2O, 99.5+ atom % D
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction