missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.1M, Chem-Lab
Click to view available options
Quantity:
5 L
Packaging:
PE
Unit Size:
5 L
Specifications
Specifications
| CAS | 7664-93-9 |
| Concentration | 9.81g H2SO4 / 1L H2O |
| Density | 1.02g/mL |
| Packaging | PE |
| Normality | 0.2N (±0.0004 at 20°C) |
| Quantity | 5 L |
| Molecular Formula | H2O4S |
| MDL Number | MFCD00064589 |
| UN Number | UN3264 |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| Show More |
Safety and Handling
Product Identifier
- Sulphuric acid 0.1 mol/l
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction