Learn More
Cole-Parmer™ Stuart™ Suction Valve For Stuart Pressmatic Bottle Top Dispenser
Click to view available options
For Use With (Equipment):
Pressmatic Bottle Top Dispenser Model PD10P
Unit Size:
Each
Description
Replacement suction valve for Stuart™ PD10P Pressmatic Bottle Top Dispenser
Specifications
Specifications
| Product Type | Suction Valve |
| For Use With (Equipment) | Pressmatic Bottle Top Dispenser Model PD10P |