missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SU 16f, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective platelet-derived growth factor receptor β (PDGFRβ) inhibitor (IC50 = 10 nM) that displays > 14-fold, > 229-fold and > 10000-fold selectivity over VEGFR2, FGFR1 and EGFR respectively. Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 = 0.11 μM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 386.44 |
Percent Purity | >98% |
Chemical Name or Material | SU 16f |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'