missing translation for 'onlineSavingsMsg'
Learn More
Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals Product Code.: 11305897

Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Product Code. 11305897
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11305897

Brand: Thermo Scientific Alfa Aesar 043667.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Styrene-d{8} is used in Organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Density 0.979 g/mL
Flash Point 31°C (88°F)
Packaging Ampule
Refractive Index 1.5445
Sensitivity Air
Quantity 5 g
Boiling Point 50°C to 51°C (25mmHg)
Linear Formula C6D5CD=CD2
UN Number UN2055
Beilstein 2327045
Solubility Information It is soluble in water 0.24g/L (20°C).
Formula Weight 112.2
Percent Purity 98%
Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.