Learn More
Waters STYRAGEL HT 6E GPC Column, THF 30cm X 7.8mm
Brand:  Waters WAT044219
Encompass
STYRAGEL HT 6E GPC Column, THF 30cm X 7.8mm
Product Code. 10005550
  • € 3345.00 / Each
Estimated Shipment: 29-10-2024