Learn More
Cole-Parmer™ Tube Holder
Click to view available options
Holds:
0.2 mLTubes
0.5 mL Tubes
1.5 mL Tubes
Unit Size:
Each
Description
- Accessory tube holders accommodate 0.2 mL or 0.5 mL microcentrifuge tubes.
- Two tube holders can be used at any one time and securely held in place by the non-slip mat.
- Stuart™ Tube Holder; For Use with Microtitre Plate Shakers where samples require controlled temperature and/or humidity conditions, ideal for placement inside CO2incubators.
Specifications
Specifications
| Holds | 1.5 mL Tubes |
| Tube Capacity | 1.5 mL |
| Product Type | Tube Holder |
| For Use With (Equipment) | Stuart™ Microtitre Plate Shakers SSM5 and SSL5 |
| Packaging Quantity | 1 Ea. |