Learn More
Techne™ Stuart™ Skirted 96-well PCR Plates
Product Code. 13038523
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 25
Product Code. 13038523
Brand: Techne™ 1012526
Only null left
Add to basket
Add to basket
These 96-well PCR plates are skirted to reduce plate distortion and provide stability.
  • Fully skirted to reduce distortion and promote stability
  • Available in Black, Clear and White
Specifications
Format Standard
Array 8 x 12
Color Clear
Product Type 96-well PCR Plate
No. of Wells 96
No. per Pack 25
Profile Low
Quantity 25 Pk.