Learn More
Cole-Parmer™ Drum for DR-200 Series Rotating Shaker System Drives
Product Code. 11456548
Click to view available options
No. of Positions:
1
2
4
240
For Use With (Equipment):
1.5 mL Microcentrifuge Tubes
250 mL Flasks or Bottles
Bottles
Clip Plates
One Bottle, Figure-8 Motion
Unit Size:
Each
Product Code. 11456548
Brand: Cole-Parmer™ 5160099
Only null left
Add to basket
Add to basket
Optimize performance of your DR-200 Series Rotating Shaker System with interchangeable drums.
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Spécification
Product Type Drum
For Use With (Application) For changing your rotator's capacity
For Use With (Equipment) 1.5 mL Microcentrifuge Tubes
No. of Positions 240