Learn More
Techne™ Stuart™ 96-well Low Profile PCR Plates
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 25
Description
- Low profile design decreases dead space between the lid of the thermal cycler, increasing PCR efficiency
- Available in Black, White and Clear
- Non-sterile
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | White |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| No. per Pack | 25 |
| Profile | Low |
| Quantity | 25 Pk. |