Learn More
Corning™ Stripwell™ Microplates
GREENER_CHOICE
Product Code. 10347891 Shop All Corning Products
Click to view available options
Color:
Black
Clear
White
Binding Property:
High Binding
Medium Binding
Total Volume (Metric) Well:
190 μL
360 μL
Unit Size:
Pack of 100
Product Code. 10347891
Brand: Corning™ 3913
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Order Info
Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.
Certifications
Cat. Nos. 07-200-31 and -32 have certified surface chemistry.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Strip Plate
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Total Volume (Metric) Well 360 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.