missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (R-PE)

Streptavidin, (R-PE)

€ 291.00 - € 1000.00

Specifications

Concentration 1 mg/mL
Conjugate R-phycoerythrin
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Content And Storage Contains 1 vial of streptavidin,R-PE conjugate.

Store in refrigerator (2–8°C).
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10503785
View Documents
Invitrogen™
SA10041
0.5 mL
€ 291.00
Each
Please to purchase this item. Need a web account? Register with us today!
10222643
View Documents
Invitrogen™
SA10044
2 mL
€ 1000.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide
Specifications

Specifications

1 mg/mL
Fluorescence
Contains 1 vial of streptavidin,R-PE conjugate.

Store in refrigerator (2–8°C).
488, 488 or 561, 561
Solution
Streptavidin Conjugate (Fluorescent)
R-phycoerythrin
Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
488/575
Wet Ice
PBS Buffer
R-PE
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Streptavidin, (R-PE)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.