missing translation for 'onlineSavingsMsg'
Learn More

Cerillo™ Stratus Plate Reader

The Stratus is small enough to fit in the palm of your hand and allows for measurements in small spaces or restrictive environments, like anaerobic or microaerophilic chambers and incubators. Its uniquely simple interface and data storage allow it to be placed anywhere, with or without a computer.

Brand:  Cerillo™ R2650

Encompass_Preferred

 View more versions of this product

Product Code. 17381819

  • € 6057.65 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Stratus is small enough to fit in the palm of your hand and allows for measurements in small spaces or restrictive environments, like anaerobic or microaerophilic chambers and incubators. Its uniquely simple interface and data storage allow it to be placed anywhere, with or without a computer.

The Stratus plate reader has new and improved capabilities to optimize flexibility for sample data collection in multiple standard plate formats to serve the wide-ranging needs of scientific researchers. The maximum allowable shaking speed is determined by the shaking radius of the shaking platform used, as well as the method of adapter attachment.

  • Available in standard 600 nm, 496 nm, 650 nm, or customized wavelength
  • Fits standard height 6-, 12-, 24-, and 96-well plates
  • Small footprint and stackable
  • No moving parts; minimal maintenance
  • Functional in a broad range of environments
  • Data is stored on a microSD card, no need for direct connection to a computer
  • Can be powered by a wall outlet, external battery pack, or computer
Specifications

Specifications

Digital Photosensor
50°C
45 sec.
Stratus
PC Software
0 to 3 OD
CE
500 mA USB Supply
microSD card, power adapter, USB cable
0 to 21%
<1% and 0.006 OD (0.000 to 2.000 OD)
5 V
650 nm
14.4 x 13.0 x 6.8 cm
Monochromatic Solid-State LEDs
6, 12, 24, and 96-well Plates
Kinetic
Single Wavelength
<1% and 0.015 OD (0.000 to 2.000 OD)
0 to 25%
Absorbance based kinetic assays
<1% (0.000 to 2.000 OD)
1 Ea.
0.005 OD
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.