Learn More
Cerillo™ Stratus Plate Reader
The Stratus is small enough to fit in the palm of your hand and allows for measurements in small spaces or restrictive environments, like anaerobic or microaerophilic chambers and incubators. Its uniquely simple interface and data storage allow it to be placed anywhere, with or without a computer.
Brand: Cerillo™ R2CUST
Description
The Stratus is small enough to fit in the palm of your hand and allows for measurements in small spaces or restrictive environments, like anaerobic or microaerophilic chambers and incubators. Its uniquely simple interface and data storage allow it to be placed anywhere, with or without a computer.
The Stratus plate reader has new and improved capabilities to optimize flexibility for sample data collection in multiple standard plate formats to serve the wide-ranging needs of scientific researchers. The maximum allowable shaking speed is determined by the shaking radius of the shaking platform used, as well as the method of adapter attachment.
- Available in standard 600 nm, 496 nm, 650 nm, or customized wavelength
- Fits standard height 6-, 12-, 24-, and 96-well plates
- Small footprint and stackable
- No moving parts; minimal maintenance
- Functional in a broad range of environments
- Data is stored on a microSD card, no need for direct connection to a computer
- Can be powered by a wall outlet, external battery pack, or computer
Specifications
Digital Photosensor | |
50°C | |
45 sec. | |
Stratus | |
PC Software | |
0 to 3 OD | |
CE | |
500 mA USB Supply | |
microSD card, power adapter, USB cable | |
0 to 21% | |
<1% and 0.006 OD (0.000 to 2.000 OD) | |
5 V | |
Custom |
14.4 x 13.0 x 6.8 cm | |
Monochromatic Solid-State LEDs | |
6, 12, 24, and 96-well Plates | |
Kinetic | |
Single Wavelength | |
<1% and 0.015 OD (0.000 to 2.000 OD) | |
0 to 25% | |
Absorbance based kinetic assays | |
<1% (0.000 to 2.000 OD) | |
1 Ea. | |
0.005 OD | |
2 Years |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.