Learn More
Buerkle™ LDPE Lid for Round Shaped Containers
Product Code. 10238540 Shop All Bürkle Products
Click to view available options
For Use With (Equipment):
200 L Round Container
25 L Round Container
30 L Round Container
35 L Round Container
50 L Round Container
Unit Size:
Each
Product Code. 10238540
Brand: Buerkle™ 34902001
Only null left
Add to basket
Add to basket
Buerkle™ LDPE Lid for Round Shaped Containers is the perfect solution to cover various sizes of round shaped containers.
  • LDPE
  • Lid for round shaped container
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lid
Color Transparent
Material LDPE
Closure Type Lid
For Use With (Equipment) 200 L Round Container