Learn More
Lenz Laborglasintrumente Stopcocks, Straight Bore, with Solid Glass Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02101806
 View more versions of this product
Product Code. 10454491
  • € 39.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12541-1
  • With solid glass plug and screwthread retaining device
  • One straight bore
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Specifications
6 mm
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers